Key Documents
20648
Supelco
Packed Column Ferrule Kit
About This Item
UNSPSC Code:
41121815
eCl@ss:
32110502
General description
Ferrule Test Kit for 1/4 in. O.D. Glass Packed Columns: 4 Supeltex M-1, 4 Supeltex M-2A, 4 Supeltex M-4 ferrules, a 6in./15 cm × 1/4 in. practice piece of glass tubing, and instructions.
Choose from one of the most recent versions:
Certificates of Analysis (COA)