Key Documents
20609
Supelco
Empty Glass GC Column, for Shimadzu Mini-GC (On-Column)
L × O.D. × I.D. 6.0 ft (1.8 m) × 5.0 mm × 3.0 mm
About This Item
UNSPSC Code:
41121800
material
glass column
technique(s)
gas chromatography (GC): suitable
L × O.D. × I.D.
6.0 ft (1.8 m) × 5.0 mm × 3.0 mm
column type
empty
compatibility
for use with Shimadzu Mini-GC
General description
GC columns are of two types; packed column and capillary column. Capillary column carries the gas through a central aperture which is unimpeded through the entire length of column. In general, capillary columns are made longer to produce more theoretical plates. This also helps in band dispersion eventually narrowing the peaks and resulting in high resolution.[1]
Silane treated empty glass column. Fittings are not included.
  • X (length of injector arm) = 7.48 in. (190 mm)
  • Y (length of detector arm) = 5.59 in. (142 mm)
  • S (span, injector to detector) = 4.72 in. (120 mm)
This column fits Shimadzu mini-GC (on-column injection).
Application
Empty column may be used for packing adsorbents for temperature-programmed desorption (TPD) instrumentation.[2]
Legal Information
Shimadzu is a trademark of Shimadzu Corporation
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
The determination of active filter aid adsorption sites by temperature-programmed desorption.
Zhu. Z. Y.
Journal of the American Oil Chemists' Society, 71, 189-194 (1994)
Mayssa H Mokalled et al.
Developmental biology, 406(2), 109-116 (2015-09-20)
Myocardin-Related Transcription Factors A and B (MRTF-A and MRTF-B) are highly homologous proteins that function as powerful coactivators of serum response factor (SRF), a ubiquitously expressed transcription factor essential for cardiac development. The SRF/MRTF complex binds to CArG boxes found
Raymond P. W. Scott
Introduction to Analytical Gas Chromatography (1997)