2050225
Inlet Liner, Splitless Type, Straight Design (unpacked)
Splitless, pkg of 25 ea
About This Item
packaging
pkg of 25 ea
L × O.D. × I.D.
74 mm × 6.3 mm × 2.0 mm
General description
- Fits Varian (1075 and 1077 Injector)
Application
Choose from one of the most recent versions:
Certificates of Analysis (COA)