2050201
Inlet Liner, Splitless Type, Straight Design (unpacked)
pkg of 1 ea
About This Item
material
glass liner
description
Inlet Liner, Splitless, glass, for Varian
straight
packaging
pkg of 1 ea
manufacturer/tradename
Varian 01-900109-05 (OEM)
Varian 03-949437-90 (OEM, instrument manufacturer′s nondeactivated liner)
L × O.D. × I.D.
74 mm × 6.3 mm × 2.0 mm
compatibility
configured for
for use with Varian 1075/1077 Injectors
technique used for Trace analytes
General description
- Fits Varian (1075 and 1077 Injector)
Application
Footnote
Choose from one of the most recent versions:
Certificates of Analysis (COA)