Key Documents
2048501
Supelco
Inlet Liner, Splitless Type, Dual-Taper Design (unpacked)
pkg of 1 ea
About This Item
UNSPSC Code:
41121800
packaging
pkg of 1 ea
L × O.D. × I.D.
78.5 mm × 6.5 mm × 4.0 mm
General description
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits Agilent (5890, 6890, and 7890)
  • Fits Finnigan (9001GCQ)
  • Fits Varian (CP-1177 Injector)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Fire Debris Analysis.
Stauffer E
Science, 249-252 (2007)
Els Van Hoeck et al.
Journal of chromatography. A, 1157(1-2), 1-9 (2007-05-08)
Stir bar sorptive extraction (SBSE) was applied to the enrichment of pyrethroids in water samples. Analysis was done by retention time-locked GC/MS in selected ion-monitoring (SIM) mode using trans-permethrin-d(6) as internal standard. Thermal desorption (TD) was performed in a classical