Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

20424-U

Sigma-Aldrich

Tubing Bender

for 1/4 in. O.D. tubing

Synonym(s):

TUBING BENDER 1/4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.35

description

Heavy Duty

Looking for similar products? Visit Product Comparison Guide

General description

The heavy duty benders easily bend aluminum, copper, or stainless steel tubing up to 180°, without kinking or splitting the tubing.

The three-in-one tool (20857) bends 1/8 in., 3/16 in., or 1/4 in. (3, 4, or 6 mm) tubing of standard wall thickness.

20422-U has a bend radius of 3/8 inch from center.
20424-U has a bend radius of 9/16 inch from center.
20857 (three-in-one tool) has a bend radius of 14.2 mm from center (pictured above).

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service