Key Documents
20422-U
Sigma-Aldrich
Tubing Bender
for 1/8 in. O.D. tubing
Synonym(s):
TUBING BENDER, 1/8
About This Item
UNSPSC Code:
41105108
NACRES:
NB.35
description
Heavy Duty
General description
The heavy duty benders easily bend aluminum, copper, or stainless steel tubing up to 180°, without kinking or splitting the tubing.
The three-in-one tool (20857) bends 1/8 in., 3/16 in., or 1/4 in. (3, 4, or 6 mm) tubing of standard wall thickness.
20422-U has a bend radius of 3/8 inch from center.
20424-U has a bend radius of 9/16 inch from center.
20857 (three-in-one tool) has a bend radius of 14.2 mm from center (pictured above).
Choose from one of the most recent versions:
Certificates of Analysis (COA)