Key Documents
20407
Supelco
O-ring for Packed Column, 1/4 in. Column O.D.
pkg of 100 ea, I.D. 1/4 in., 1/4 in. O.D. Column
About This Item
UNSPSC Code:
41121815
description
packed
packaging
pkg of 100 ea
maximum temperature
200 °C
technique(s)
gas chromatography (GC): suitable
I.D.
1/4 in.
compatibility
1/4 in. O.D. Column
General description
O-ring sealing components are required for machines that need to seal fluids or gases.[1] Column end piece tend to trap liquid around the O-ring sealing, to avoid such troubleshoot the upper column end piece can be redesigned.[2]
Application
O-ring maybe used to prevent liquid channelling along the wall of the packed column in the experiment investigating the absorption and desorption of CO2 in a packed column.[3]
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Sofronas A.
Analytical Troubleshooting of Process Machinery and Pressure Vessels (2006)
Peptide synthesis. Part 13. Feedback control in solid phase synthesis. Use of fluorenylmethoxycarbonyl amino acid 3, 4-dihydro-4-oxo-1, 2, 3-benzotriazin-3-yl esters in a fully automated system.
Journal of the Chemical Society, 1 (10), 2895-2901 (1988)
Study of CO2 absorption and desorption in a packed column.
James TY
Energy and Fuels, 15 (2), 274-278 (2001)