Key Documents
20402
Dremel Engraver
AC/DC input 110 V AC
About This Item
UNSPSC Code:
41121800
General description
Eliminate gaps when packing your columns. The Burgess Vibrograver is ideal for vibrating columns or marking tags.
  • 110 VAC models: 9 watts, 0.08 amp, 60Hz.
  • 220 VAC model: 12 watts, 0.05 amp, 50Hz; not CE compliant.
Choose from one of the most recent versions:
Certificates of Analysis (COA)