Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

20379

Supelco

Carbotrap® 300 Dynatherm 890, 850

pkg of 1 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008

material

glass TD tube (non-fritted)

agency

EPA TO-1,TO-2,TO-3,IP-1B,TO-17

feature

preconditioned

composition

Carbosieve S-III
Carbotrap B
Carbotrap C

packaging

pkg of 1 ea

manufacturer/tradename

Supelco Carbotrap® 300

I.D. × O.D.

4 mm × 6 mm

matrix

Carbotrap® 300

application(s)

environmental
industrial hygiene

compatibility

for use with Dynatherm ACEM-9300, ACEM-900 850/890 TDU, MTDU and OI Analytical Air Tube Desorber or DMP-16

Application

C2 and larger compounds in air

Legal Information

Carbotrap is a registered trademark of Merck KGaA, Darmstadt, Germany
Dynatherm is a trademark of CDS Analytical

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Franziska Grzegorzewski et al.
Polymers, 11(9) (2019-09-13)
A simple and effective way to prepare multi-walled carbon nanotubes (MWNT)//silica hybrid microcapsules (colloidosomes) is presented. These microcapsules have been generated by emulsion templating in a biphasic oil-in-water (o/w) system. Two trialkoxysilanes of complementary polarity, (3-aminopropyl)triethoxysilane (APTES) and dodecyltriethoxysilane (DTES)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service