Key Documents
20353
Septum Puller
screw design
About This Item
UNSPSC Code:
41121800
packaging
pkg of 1 ea
technique(s)
gas chromatography (GC): suitable
L
4 in.
General description
The screw septum puller is perfect for removing harder, high temperature septa from injection ports.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
If you need assistance, please contact Customer Support
Already Own This Product?
Visit the Document Library
Vikram Babu Kasaragod et al.
Neuron, 101(4), 673-689 (2019-02-02)
The frontline anti-malarial drug artemisinin and its derivatives have also been implicated in modulating multiple mammalian cellular pathways, including the recent identification of targeting γ-aminobutyric acid type A receptor (GABAAR) signaling in the pancreas. Their molecular mechanism of action, however