Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

20346

Porapak Porous Polymer Adsorbent

matrix Porapak PS, 80-100 mesh, bottle of 75 cc

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100

form

solid

Quality Level

packaging

bottle of 75 cc

parameter

250 °C temp. limit

technique(s)

gas chromatography (GC): suitable

surface area

~150 m2/g

matrix

Porapak PS

particle size

80-100 mesh

density

~0.26 g/mL (free fall density)

Looking for similar products? Visit Product Comparison Guide

General description

Porapak porous polymers are spherical beads with consistent particle size, porosity, and surface area that help to insure analytical precision and accuracy.

For more information about any of our adsorbents, please visit sigma-aldrich.com/adsorbents
Porous polymers are the most suitable adsorbent for applications based on the analysis of gases, acids, amines, organics of low carbon number and water.[1] They are best suited for gas chromatography and are basically copolymers of polydivinylbenzene (DVB) which are very porous in nature ranging from mesoporous to microporous.[2] Porapak PS is suitable for the separation of esters, ethers, ketones, alcohols, hydrocarbons, fatty acids, aldehydes and glycols.[3]

Features and Benefits

Porapak PS is a silanized version of Porapak P to reduce surface adsorption.

Legal Information

PoraPak is a trademark of Waters

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Contemporary Practice of Chromatography.
Poole CF and Schuette SA.
Science, 70-71 (1984)
Grob RL and Barry EF.
Modern Practice of Gas Chromatography, 79-83 (2004)
Poole C.
Gas Chromatography, 127-130 (2012)

Questions

1–6 of 6 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. How can I purchase a column that is packed with this material?

    1 answer
    1. Most packed column are custom made and require that you obtain a quotation. Please contact our Customer & Sales Services group at phone number (800)-247-6628, fax number (800)-447-3044 or email [email protected] for additional information.

      Helpful?

  3. What does the 80/100 stand for in the product description?

    1 answer
    1. The 80/100 refers to the mesh size range of the actual particles.  When working with mesh size: the larger the mesh number, the smaller the diameter of the particle.  In this case 80 refers to a diameter of 177 microns and 100 refers to a diameter of 149 microns.  Therefore; the particles in this product with 80/100 will range between 149 microns and 177 microns.

      Helpful?

  4. What is the weight in grams of the 75cc HayeSep® container?

    1 answer
    1. The weight is approximately 24 grams for each 75cc container.

      Helpful?

  5. What is the maximum temperature for this packing?

    1 answer
    1. The maximum temperature for this packing is 250 °C.

      Helpful?

  6. Is this product a packed column?

    1 answer
    1. This is packing material only; it is not a column. Some endusers will purchase this bottle of packing and prepare their own packed columns.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service