Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

20335

Porapak Porous Polymer Adsorbent

matrix Porapak R, 100-120 mesh, bottle of 75 cc

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100

form

solid

packaging

bottle of 75 cc

parameter

250 °C temp. limit

technique(s)

gas chromatography (GC): suitable

surface area

~525 m2/g

matrix

Porapak R

particle size

100-120 mesh

density

~0.32 g/mL (free fall density)

Looking for similar products? Visit Product Comparison Guide

General description

Porapak porous polymers are spherical beads with consistent particle size, porosity, and surface area that help to insure analytical precision and accuracy.

For more information about any of our adsorbents, please visit sigma-aldrich.com/adsorbents
Porous polymers are the most suitable adsorbent for applications based on the analysis of gases, acids, amines, organics of low carbon number and water.[1] They are best suited for gas chromatography and are basically copolymers of polydivinylbenzene (DVB) which are very porous in nature ranging from mesoporous to microporous.[2] Porapak R is a porous polymer composed of vinyl pyrollidone mainly suitable for the separation of ethers and esters.[1]

Application

Porapak R with Amberlite XAD-4 maybe useful for chromatographic sampling of polar and nonpolar low-volatility compounds.[3]

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.
PoraPak is a trademark of Waters

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poole C.
Gas Chromatography, 127-130 (2012)
Chromatographic characterization of porous polymers for use as adsorbents in sampling columns.
Butler LD and Burke MF.
Journal of Chromatographic Science, 14 (3), 117-122 (1976)
Grob RL and Barry EF.
Modern Practice of Gas Chromatography, 79-83 (2004)

Questions

1–6 of 6 Questions  
  1. How can I purchase a column that is packed with this material?

    1 answer
    1. Most packed column are custom made and require that you obtain a quotation. Please contact our Customer & Sales Services group at phone number (800)-247-6628, fax number (800)-447-3044 or email [email protected] for additional information.

      Helpful?

  2. Is this product a packed column?

    1 answer
    1. This is packing material only; it is not a column. Some endusers will purchase this bottle of packing and prepare their own packed columns.

      Helpful?

  3. What is the weight in grams of the 75cc HayeSep® container?

    1 answer
    1. The weight is approximately 24 grams for each 75cc container.

      Helpful?

  4. What is the maximum temperature for this packing?

    1 answer
    1. The maximum temperature for this packing is 250 °C.

      Helpful?

  5. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  6. What does the 100/120 stand for in the product description?

    1 answer
    1. The 100/120 refers to the mesh size range of the actual particles.  When working with mesh size: the larger the mesh number, the smaller the diameter of the particle.  In this case 100 refers to a diameter of 149 microns and 120 refers to a diameter of 125 microns.  Therefore; the particles in this product with 100/120 will range between 125 microns and 149 microns.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service