The maximum temperature for this packing is 400 °C.
20304
Molecular Sieve Adsorbent
matrix Molecular Sieve 13X, 45-60 mesh, bottle of 50 g
About This Item
Recommended Products
form
solid
packaging
bottle of 50 g
matrix
Molecular Sieve 13X
particle size
45-60 mesh
pore size
13 Å pore diameter
density
~0.65 g/mL (free fall density)
Looking for similar products? Visit Product Comparison Guide
General description
For more information about any of our adsorbents, please visit sigma-aldrich.com/adsorbents
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
What is the maximum temperature for this packing?
1 answer-
Helpful?
-
-
What is the pore size of Molecular Sieve Adsorbent 13X, Product 20304?
1 answer-
The pore size is 10 Angstrom for product number 20304.
Helpful?
-
-
What are some common applications for Molecular Sieve Adsorbent 13X, Product 20304?
1 answer-
Type 13X is used commercially for general gas drying, air plant feed purification (simultaneous removal of H20 and CO2) and liquid hydrocarbon and natural gas sweetening (H2S and mercapstan removal). All molecules which can be adsorbed on molecular sieves 3A, 4A and 5A can be adsorbed on Type 13X. In addition, Type 13X can adsorb molecules of larger critical diameters, such as aromatics and branched-chain hydrocarbons.
Helpful?
-
-
Is this product a packed column?
1 answer-
This is packing material only; it is not a column. Some endusers will purchase this bottle of packing and prepare their own packed columns.
Helpful?
-
-
What is the Department of Transportation shipping information for this product?
1 answer-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Helpful?
-
-
How can I purchase a column that is packed with this material?
1 answer-
Most packed column are custom made and require that you obtain a quotation. Please contact our Customer & Sales Services group at phone number (800)-247-6628, fax number (800)-447-3044 or email [email protected] for additional information.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service