Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

20207

Supelco

Chromosorb® P

AW-DMCS, 80-100 mesh particle size, bottle of 100 g

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
23151817
PubChem Substance ID:

Quality Level

packaging

bottle of 100 g

technique(s)

gas chromatography (GC): suitable

surface area

4-6 m2/g

particle size

80-100 mesh

density

0.32-0.38 g/mL at 25 °C
0.38 g/cm3 (loose weight)(lit.)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chromosorb® P diatomite packed column supports are particularly well suited for nonpolar compounds and must be deactivated for use with polar compounds. They can support high phase loadings, up to 35% by weight in some cases.

Legal Information

Chromosorb is a registered trademark of Imerys Minerals California, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrej M Kielbassa et al.
PloS one, 15(1), e0228249-e0228249 (2020-01-29)
This ex vivo proof-of-concept study aimed to investigate the effect of nanosilver particles (AgNP) added to a conventional infiltrant resin (Icon) on external penetration into natural proximal enamel caries exceeding into dentin after internal tunnel preparation and internal infiltration. Carious
Miriam Ayuso et al.
PloS one, 15(6), e0233910-e0233910 (2020-06-06)
Farmers face difficulties in redeeming their investment in larger litter sizes since this comes with larger litter heterogenicity, lower litter resilience and risk of higher mortality. Dietary oligosaccharides, given to the sow, proved beneficial for the offspring's performance. However, giving

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service