Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

20193

Supelco

Chromosorb® P

NAW, 60-80 mesh particle size, bottle of 100 g

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
PubChem Substance ID:

packaging

bottle of 100 g

technique(s)

gas chromatography (GC): suitable

particle size

60-80 mesh

density

0.32-0.38 g/mL at 25 °C
0.38 g/cm3 (loose weight)(lit.)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Chromosorb is a registered trademark of Imerys Minerals California, Inc.

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

STOT RE 1 Inhalation

target_organs

Lungs

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leah H Cobb et al.
PloS one, 14(11), e0220421-e0220421 (2019-11-23)
Osteomyelitis, or bone infection, is often induced by antibiotic resistant Staphylococcus aureus strains of bacteria. Although debridement and long-term administration of antibiotics are the gold standard for osteomyelitis treatment, the increase in prevalence of antibiotic resistant bacterial strains limits the
María Jesús González-Pabón et al.
PloS one, 14(9), e0222538-e0222538 (2019-10-01)
Microbial fuel cells (MFCs) can evolve in a viable technology if environmentally sound materials are developed and became available at low cost for these devices. This is especially important not only for the designing of large wastewater treatment systems, but
Mika Azaki et al.
PloS one, 14(11), e0225177-e0225177 (2019-11-15)
Francisella tularensis (F. tularensis) is highly pathogenic to humans and must be handled under biosafety level 3 conditions. Samples used for the diagnosis and experimental analysis must be completely inactivated, although methods for the inactivation of F. tularensis are limited.
Freddy Magdama et al.
PloS one, 14(9), e0222727-e0222727 (2019-09-24)
Fusarium oxysporum f. sp. cubense Tropical Race 4 (Foc TR4) is threatening banana production worldwide. Despite quarantine efforts, the pathogen continues to spread; thus, early diagnosis plays an essential role for the proper execution of contingency plans. Here, we assess
Larysa V Yuzefovych et al.
PloS one, 14(10), e0222278-e0222278 (2019-10-11)
Cells damaged by mechanical or infectious injury release proinflammatory mitochondrial DNA (mtDNA) fragments into the circulation. We evaluated the relation between plasma levels of mtDNA fragments in obese type 2 diabetes mellitus (T2DM) patients and measures of chronic inflammation and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service