Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

20047

Supelco

ORBO 356 4-TBC on Activated Coconut Charcoal (20/40), 400/200 mg

W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea

Synonym(s):

TBC on Charcoal Tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131900

material

W,W,W separators

agency

OSHA 89,92,94

product line

ORBO

composition

Bed A, 400 mg
Bed B, 200 mg

packaging

pkg of 50 ea

manufacturer/tradename

ORBO 356

technique(s)

active air sampling: suitable

O.D. × L

8 mm × 110 mm

matrix

4-tert-butyl catechol (4-TBC) on charcoal
charcoal

matrix active group

4-tert-butylcatechol

application(s)

air monitoring
environmental
industrial hygiene

compatibility

mode of use Solvent desorption
technique used for Air monitoring

Looking for similar products? Visit Product Comparison Guide

Legal Information

ORBO is a trademark of Sigma-Aldrich Co. LLC

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service