20041
ORBO™ 304 Charcoal low nickel content (20/40), 120/60 mg
W,F,F separators, O.D. × L 6 mm × 80 mm, pkg of 50 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
EC Number:
UNSPSC Code:
47131900
Recommended Products
material
W,F,F separators
agency
NIOSH 6007
product line
ORBO™
composition
Bed A, 120 mg
Bed B, 60 mg
packaging
pkg of 50 ea
manufacturer/tradename
ORBO™ 304
technique(s)
active air sampling: suitable
O.D. × L
6 mm × 80 mm
matrix
Charcoal (low Ni)
particle size
20-40 mesh
application(s)
air monitoring
environmental
industrial hygiene
General description
ORBO sorbent tubes contain two beds of the same selective adsorbent separated by glass wool or foam, for gas and vapor sampling. The dual-layer or two-bed construction of the tube allows for any sample breakthrough to be captured in the smaller backup bed.
Legal Information
ORBO is a trademark of Sigma-Aldrich Co. LLC
Storage Class
13 - Non Combustible Solids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service