Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

18183

Supelco

(+)-Pulegone

reference material

Synonym(s):

(R)-2-Isopropylidene-5-methylcyclohexanone, (R)-p-Menth-4(8)-en-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Beilstein/REAXYS Number:
2040703
MDL number:
UNSPSC Code:
41116107

grade

reference material

assay

≥97.0% (GC)

form

liquid

optical activity

[α]/D 23.0±2.0°, neat

shelf life

limited shelf life, expiry date on the label

bp

223-224 °C (lit.)

density

0.935 g/mL at 20 °C (lit.)

storage temp.

−20°C

SMILES string

C[C@@H]1CCC(=C(\C)C)\C(=O)C1

InChI

1S/C10H16O/c1-7(2)9-5-4-8(3)6-10(9)11/h8H,4-6H2,1-3H3/t8-/m1/s1

InChI key

NZGWDASTMWDZIW-MRVPVSSYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

pictograms

Health hazardExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

179.6 °F - closed cup

flash_point_c

82 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service