Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

150693

Supelco

Cerium(IV) sulfate solution

0.1 M, for 3S adapter technology

Synonym(s):

Ceric sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ce(SO4)2
CAS Number:
Molecular Weight:
332.24
MDL number:
UNSPSC Code:
12352302

form

liquid

reaction suitability

reaction type: Redox Reactions

concentration

0.0995-0.1005 M
0.1 M

technique(s)

titration: suitable

volume

1 L
1000 mL

SMILES string

[Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/Ce.2H2O4S/c;2*1-5(2,3)4/h;2*(H2,1,2,3,4)/q+4;;/p-4

InChI key

VZDYWEUILIUIDF-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

3S - Safe, Smart, Secure
An intelligent packaging solution protected with originality seal.
Reagent information is transferred via RFID from reagent bottle to titrator.
3S reagents are compatible only with the Metrohm Omnis titration platform.

Please see here SigmaAldrich.com/3S

Other Notes

Once the bottle is opened, storage time is limited.

pictograms

CorrosionEnvironment

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service