Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

150689

Supelco

Silver nitrate solution

0.1 M, for 3S adapter technology

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
AgNO3
CAS Number:
Molecular Weight:
169.87
MDL number:
UNSPSC Code:
12352302

form

liquid

reaction suitability

reaction type: Precipitation reactions

concentration

0.0995-0.1005 M
0.1 M

technique(s)

titration: suitable

volume

1 L
1000 mL

SMILES string

[Ag]

InChI

1S/Ag.NO3/c;2-1(3)4/q+1;-1

InChI key

SQGYOTSLMSWVJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3S - Safe, Smart, Secure
An intelligent packaging solution protected with originality seal.
Reagent information is transferred via RFID from reagent bottle to titrator.
3S reagents are compatible only with the Metrohm Omnis titration platform.

Please see here SigmaAldrich.com/3S

Other Notes

Once the bottle is opened, storage time is limited.

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Met. Corr. 1 - Repr. 1B - Skin Irrit. 2

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service