150677
Hydrochloric acid solution
0.1 M, for 3S adapter technology
Synonym(s):
Hydrogen chloride solution
About This Item
Recommended Products
vapor pressure
190 mmHg ( 25 °C)
form
liquid
reaction suitability
reaction type: Acid-base reactions
concentration
0.0995-0.1005 M
0.1 M
technique(s)
titration: suitable
volume
1 L
1000 mL
bp
100 °C (lit.)
density
1 g/cm3
SMILES string
Cl
InChI
1S/ClH/h1H
InChI key
VEXZGXHMUGYJMC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
An intelligent packaging solution protected with originality seal.
Reagent information is transferred via RFID from reagent bottle to titrator.
3S reagents are compatible only with the Metrohm Omnis titration platform.
Please see here SigmaAldrich.com/3S
Other Notes
signalword
Warning
hcodes
pcodes
Hazard Classifications
Met. Corr. 1
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service