13976-U
Metal Packed GC Column (General Configuration)
phase 30% DC-200 (500 cstks), matrix 80/100 Chromosorb P NAW support, L × O.D. × I.D. 1.5 ft (0.46 m) × 1/8 in. × 2.1 mm
About This Item
Recommended Products
material
stainless steel column
technique(s)
gas chromatography (GC): suitable
L × O.D. × I.D.
1.5 ft (0.46 m) × 1/8 in. × 2.1 mm
matrix
80/100 Chromosorb P NAW support
matrix active group
30% DC-200 (500 cstks) phase
column type
packed GC
compatibility
for use with General Configuration
General description
signalword
Danger
hcodes
Hazard Classifications
STOT RE 1 Inhalation
target_organs
Lungs
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 1
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service