Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13678

Supelco

Dowex Optipore® V-503

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
23151817

form

solid

packaging

bucket of 1 kg

technique(s)

LPLC: suitable
thin layer chromatography (TLC): suitable

ash

<0.01%

color

orange-brown

matrix

styrene-divinylbenzene

matrix active group

polymer

particle size

1 mm

pore size

0.94 mL/g pore volume
34 Å mean pore size

bulk density

340 g/L (9.66 kg/ft3)

separation technique

reversed phase

Application

Dowex Optipore® V-503 may be used in separation medium for TLC, paper chromatography and LPLC.
Polymeric Adsorbent for Removal of Organics from Humid Air Streams.
DOWEX OPTIPORE V503 is a 1.0 mm diameter, spherical bead material, designed to give lower pressure drop in vapor phase applications while retaining all the other attributes of the smaller particle size adsorbent, DOWEX OPTIPORE V493.
In contrast to activated carbon, DOWEX OPTIPORE V503 adsorbent can be used to adsorb reactive solvents without catalyzing their decomposition, due to its extremely low mineral ash content.
Because DOWEX OPTIPORE V503 is a powerful adsorbent, it may adsorb odors and solvents during transportation and storage. The adsorbent can be precycled through a regeneration cycle prior to use to remove these materials.

Other Notes

For development of new applications, use the DOWEX OPTIPORE® V-503. Inquire for pricing and availability.

Legal Information

DOWEX OPTIPORE is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service