Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

13563

Supelco

Diaion® PK228L sodium form

pkg of 1000 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817

description

cross-linkage 14%

form

beads

packaging

pkg of 1000 g

parameter

120 °C max. temp.

moisture

37-43%

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (porous)

matrix active group

SCX

particle size

16-50 mesh

operating pH

0-14

capacity

2.0 meq/mL by wetted bed volume
4.2 meq/g by dry weight

separation technique

cation exchange

Application

Diaion® PK228L sodium form may be used in separation medium for TLC, paper chromatography and LPLC.
Strongly acidic cation exchange resin for decolorization, softening and deionization of water, recovery and separation of metals, refining of chemicals, sugar and dextrose, catalysis, and separation of amino acids. The resin has good stability against organic fouling and oxidation, and meets requirements of FDA Food Additive Regulation 21 CFR 173.25.

Legal Information

Diaion is a registered trademark of Mitsubishi Chemical Corp.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service