13122-U
Metal Packed GC Column (fits Agilent, Configuration "A")
phase 10% SE-30, matrix 80/100 Chromosorb W HP support, L × O.D. × I.D. 6.0 ft (1.8 m) × 1/8 in. × 2.1 mm
About This Item
Recommended Products
material
stainless steel column
technique(s)
gas chromatography (GC): suitable
L × O.D. × I.D.
6.0 ft (1.8 m) × 1/8 in. × 2.1 mm
matrix
80/100 Chromosorb W HP support
matrix active group
10% SE-30 phase
column type
packed GC
compatibility
for use with Agilent Configuration A
Looking for similar products? Visit Product Comparison Guide
General description
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service