Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12606

Supelco

Silica gel on TLC Al foils

L × W 5 cm × 20 cm, with fluorescence indicator 254 nm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

material

aluminum support
silica gel 60 matrix

feature

binder Polymeric
fluorescent indicator

manufacturer/tradename

Sigma-Aldrich

technique(s)

thin layer chromatography (TLC): suitable

L × W

5 cm × 20 cm

layer thickness

200 μm

particle size

8.0-12.0 μm

pore size

0.75 mL/g pore volume
60 Å mean pore size

Looking for similar products? Visit Product Comparison Guide

General description

  • Outstanding wettability (even for 100% aqueous visualisation reagents)
  • Easy and precise cutting (no flaking of silica)
  • Excellent separation efficiency and reproducibility from batch to batch
Silica gel, highly porous textured matrix are generally used for synthesis of adsorbents. They are corpuscular in structure. Silica surface possess a layer of hydroxyl groups and adsorbed water molecules, which can be removed by drying at high temperature or even by treatment with suitable reagents. Silica gel surface plays important role in catalysis, chemical separation and microelectronic fabrications. Aluminium sheets precoated with silica gel, are used as stationary phase in TLC.

Application

TLC Al foils may be used for Thin layer chromatography.

Other Notes

this product will be discontinued, please find the alternative product here :
105570

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Estimation of Gemfibrozil in tablet dosage form by HPTLC method.
Vikas Parikh C and Karkhanis VV.
Research Journal of Pharmaceutical, Biological and Chemical Sciences, 2, 889-898 (2011)
MODIFIED AND UNMODIFIED SILICA GEL USED FOR HEAVY METAL IONS REMOVAL FROM AQUEOUS SOLUTIONS
Tzvetkova, P., and R. Nickolov.
Journal of the University of Chemical Technology and Metallurgy, 47, 498-504 (2012)
Proton NMR study of dehydration of the silica gel surface
Bronnimann
Journal of the American Chemical Society, 110, 2023-2026 (1988)
The surface structure of silica gel
Peri, J. B., and A. L. Hensley Jr.
The Journal of Physical Chemistry, 72, 2926-2933 (1968)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service