12489
Metal Packed GC Column (fits Varian®)
phase SP®-1000, 1%, matrix 60/80 Carbopack B support, L × O.D. × I.D. 6.0 ft (1.8 m) × 1/8 in. × 2.1 mm
About This Item
material
stainless steel column
manufacturer/tradename
Varian
parameter
0-225 °C temperature (isothermal or programmed)
technique(s)
gas chromatography (GC): suitable
L × O.D. × I.D.
6.0 ft (1.8 m) × 1/8 in. × 2.1 mm
matrix
60/80 Carbopack B support
matrix active group
SP®-1000 phase, 1%
column type
packed GC
compatibility
for use with Varian
Looking for similar products? Visit Product Comparison Guide
General description
Application
Legal Information
Storage Class
11 - Combustible Solids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service