12481-U
Glass Packed GC Column (fits Agilent, Configuration "A")
phase none, matrix 100/120 Porapak S support, L × O.D. × I.D. 6.6 ft (2.0 m) × 1/4 in. × 4.0 mm, Preconditioned
About This Item
material
glass column
manufacturer/tradename
Agilent
parameter
0-250 °C temperature (isothermal or programmed)
technique(s)
gas chromatography (GC): suitable
L × O.D. × I.D.
6.6 ft (2.0 m) × 1/4 in. × 4.0 mm, Preconditioned
matrix
100/120 Porapak S support
matrix active group
none phase
column type
packed GC
compatibility
for use with Agilent Configuration A
Looking for similar products? Visit Product Comparison Guide
General description
Other Notes
This column is preconditioned.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service