Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

12384

Supelco

Metal Packed GC Column (fits Varian®)

phase none, matrix 40/60 Carboxen-1000 support, L × O.D. × I.D. 5.0 ft (1.5 m) × 1/8 in. × 2.1 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115710
eCl@ss:
32110502

material

stainless steel column

manufacturer/tradename

Varian

parameter

0-225 °C temperature (isothermal or programmed)

technique(s)

gas chromatography (GC): suitable

L × O.D. × I.D.

5.0 ft (1.5 m) × 1/8 in. × 2.1 mm

matrix

40/60 Carboxen-1000 support

matrix active group

none phase

column type

packed GC

compatibility

for use with Varian

Looking for similar products? Visit Product Comparison Guide

General description

An economical metal column should be used for less demanding applications where the inertness of glass is not required. This column fits Varian 3000, 4000, 6000, and Vista (FID, inj A to det A).
Packed column made up of stainless steel are basically employed when higher carrier gas flow rates are employed as they can sustain elevated pressure. Stainless steel columns are usually washed with dilute hydrochloric acid, then with water followed by methanol, acetone, methylene dichloride and n-hexane in order to remove any corrosion products and traces of lubricating agents.[1]

Application

Carboxen 1000 packed in a stainless steel column maybe used in the anaerobic chamber for conductance of research with methanogenic archaea.[2]

Legal Information

Varian is a registered trademark of Varian Medical Systems, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Archaea: A Laboratory Manual: Methanogens.
Robb FT, et al.
Science, 28-30 (1994)
Qingshan Shen et al.
Carbohydrate polymers, 222, 115015-115015 (2019-07-20)
Co-production of chondroitin sulfate (CS) and peptides was realized from the liquid fraction of chicken sternal cartilage subjected to hot-pressure (HP) by membrane combination separation technology. Cartilage was liquefied via the HP treatment at 110 °C (0.07 MPa) and 120 °C (0.1 MPa) for
Introduction to Analytical Gas Chromatography.
Scott RPW.
Science, 102-108 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service