The maximum temperature for this packing is 275 °C.
11833
Coated GC Packing
phase 3% SP®-2310 + 2% {tm="SP"]-2300, matrix 100/120 Chromosorb W AW support, bottle of 20 g
About This Item
Recommended Products
packaging
bottle of 20 g
parameter
25-275 °C temp. range
technique(s)
gas chromatography (GC): suitable
matrix
100/120 Chromosorb W AW support
matrix active group
3% SP®-2310 + 2% {tm="SP"]-2300 phase
column type
packed GC
compatibility
for analyte group Rapeseed FAME′s
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
signalword
Danger
hcodes
Hazard Classifications
STOT RE 1 Inhalation
target_organs
Lungs
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
What is the maximum temperature for this packing?
1 answer-
Helpful?
-
-
What does the 100/120 stand for in the product description?
1 answer-
The 100/120 refers to the mesh size range of the actual particles. When working with mesh size: the larger the mesh number, the smaller the diameter of the particle. In this case 100 refers to a diameter of 149 microns and 120 refers to a diameter of 125 microns. Therefore; the particles in this product with 100/120 will range between 125 microns and 149 microns.
Helpful?
-
-
Is this product a packed column?
1 answer-
This is packing material only; it is not a column. Some endusers will purchase this bottle of packing and prepare their own packed columns.
Helpful?
-
-
What is the Department of Transportation shipping information for this product?
1 answer-
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
Helpful?
-
-
How can I purchase a column that is packed with this material?
1 answer-
Most packed column are custom made and require that you obtain a quotation. Please contact our Customer & Sales Services group at phone number (800)-247-6628, fax number (800)-447-3044 or email [email protected] for additional information.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service