11060
GC Stationary Phase
phase Ethylene glycol adipate (EGA), bottle of 25 g
About This Item
Recommended Products
agency
meets requirements for USP G23
form
solid
packaging
bottle of 25 g
parameter
100-225 °C temp. range
technique(s)
gas chromatography (GC): suitable
matrix active group
Ethylene glycol adipate (EGA) phase
solubility
acetone: soluble
column type
packed GC
General description
Application
Kit Components Only
- Di[poly(ethylene glycol)] adipate
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service