Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11033-U

Supelco

GC Stationary Phase

phase Carbowax 20M/terephthalic acid, bottle of 50 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
SB.54

agency

meets requirements for USP G25

form

solid

packaging

bottle of 50 g

parameter

60-225 °C temp. range

technique(s)

gas chromatography (GC): suitable

matrix active group

Carbowax 20M/terephthalic acid phase

solubility

chloroform: soluble

column type

packed GC

Looking for similar products? Visit Product Comparison Guide

General description

Synthesized specifically to be purer, of narrow molecular weight range, and without trace catalysts or impurities for use as a GC stationary phase.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service