Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

10389

Supelco

Amberlyst® 15 hydrogen form

dry

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817

form

beads

quality

dry

packaging

pkg of 100 g

parameter

120 °C max. temp.

moisture

≤1.6%

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (macroreticular)

matrix active group

SCX

particle size

<300 μm

operating pH

0-14

capacity

1.7 meq/mL by wetted bed volume
4.7 meq/g by dry weight

separation technique

cation exchange

InChI

1S/C10H10.C8H8O3S/c1-3-9-7-5-6-8-10(9)4-2;1-2-7-5-3-4-6-8(7)12(9,10)11/h3-8H,1-2H2;2-6H,1H2,(H,9,10,11)

InChI key

SIWVGXQOXWGJCI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Amberlyst 15, a macro reticular polystyrene based ion exchange resin with strongly acidic sulfonic group, serves as an excellent source of strong acid. It can also be used several times.[1] Amberlyst 15 is a heat sensitive resin which can be used as a catalyst.[2]

Application

Amberlyst® 15 may be used:
  • as column packing material for HPLC analysis
  • in separation media for TLC, paper and LPL chromatography
Strongly acidic cation exchange resin used as a heterogeneous acid catalysis; suitable for non-aqueous catalysis.

Chem/Phys Resistance

Swelling (dry to solvent saturated, %):
hexane 10-15; toluene 10-15; ethylene dichloride 15-20; ethylene acetate 30-40; 95% ethylene alcohol 60-70; water 60-70.

Legal Information

Amberlyst is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amberlyst-15 in organic synthesis
Rammohan Pal,et al.
ARKIVOC (Gainesville, FL, United States), i, 570-609 (2012)
Determination of adsorption and kinetic parameters for methyl acetate esterification and hydrolysis reaction catalyzed by Amberlyst 15
Yu, Weifang, K. Hidajat, and Ajay K. Ray
Applied Catalysis A: General, 260, 191-205 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service