Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

10353

Supelco

Duolite® C467 sodium form

bucket of 500 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817

packaging

bucket of 500 g

parameter

65 °C max. temp.

moisture

65-72%

matrix

polymeric (macroreticular)

matrix active group

functional group, aminophosphonic

particle size

16-50 mesh

operating pH

1-10

capacity

1.0 meq/mL by wetted bed volume
3.5 meq/g by dry weight

Analysis Note

Chelating resin especially useful for capturing metallic cations of low mass, removing of hardness from brine, and as a catalyst (resistant to osmotic shock).

Legal Information

Duolite is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service