Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07346

Supelco

Cyromazin

reference material

Synonym(s):

N-Cyclopropyl-1,3,5-triazin-2,4,6-triamine, Cyromazin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10N6
CAS Number:
Molecular Weight:
166.18
Beilstein/REAXYS Number:
882879
MDL number:
UNSPSC Code:
41116107

grade

reference material

assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

mp

223-227 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

Nc1nc(N)nc(NC2CC2)n1

InChI

1S/C6H10N6/c7-4-10-5(8)12-6(11-4)9-3-1-2-3/h3H,1-2H2,(H5,7,8,9,10,11,12)

InChI key

LVQDKIWDGQRHTE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyromazin, an insect growth regulator, belongs to the class of triazine pesticides. It is widely used in agriculture for protecting fruits, vegetables, and flowers.[1] This insecticide primarily affects the insect cuticle. On a large scale, it is found to be effective against fly larvae and leaf miners. It also helps to control insect flies in cattle manure and agriculture.[2][3]
This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Application

Cyromazin may be used as reference material for the quantification of the analyte in milk/milk powder,[1] chicken eggs,[4] and pesticide formulations[3] using chromatography techniques.

pictograms

Environment

hcodes

Hazard Classifications

Aquatic Chronic 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

212.0 °F - closed cup

flash_point_c

100 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Disposition of cyromazine in plants under environmental conditions
Lim OL, et al.
Journal of Agricultural and Food Chemistry, 38(3), 860-864 (1990)
A novel dispersive micro solid phase extraction using PCX as the sorbent for the determination of melamine and cyromazine in milk and milk powder by UHPLC-HRMS/MS
Chen D, et al.
Talanta, 134, 144-152 (2015)
Determination of cyromazine in pesticide commercial formulations by vibrational spectrometric procedures
Armenta S, et al.
Analytica Chimica Acta, 524(1-2), 257-264 (2004)
Pei-Cheng Wang et al.
Analytica chimica acta, 752, 78-86 (2012-10-30)
A rapid and sensitive method has been developed for the simultaneous detection of cyromazine and melamine in chicken eggs using the quick, easy, cheap, effective, rugged and safe (QuEChERS) method coupled with liquid chromatography-tandem mass spectrometry (LC-MS/MS). The optimal extraction

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service