Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

06721

Supelco

Benzoylprop-ethyl

reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H17Cl2NO3
CAS Number:
Molecular Weight:
366.24
Beilstein/REAXYS Number:
2899940
MDL number:
UNSPSC Code:
41116107

grade

reference material

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

format

neat

SMILES string

CCOC(=O)C(C)N(C(=O)c1ccccc1)c2ccc(Cl)c(Cl)c2

InChI

1S/C18H17Cl2NO3/c1-3-24-18(23)12(2)21(14-9-10-15(19)16(20)11-14)17(22)13-7-5-4-6-8-13/h4-12H,3H2,1-2H3

InChI key

SLCGUGMPSUYJAY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzoylprop-ethyl is a post-emergence selective herbicide belonging to the dichloroaniline propionate family. Its mode of action involves the inhibition of lipid biosynthesis. This herbicide is commonly used for the control of wild oats and wild barley in winter wheat.[1][2]
This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Application

Benzoylprop-ethyl may be used as a reference material for the quantification of the analyte in Chinese teas,[3] fruits, and vegetables[4] using chromatography techniques.

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The fate of the herbicide benzoylprop-ethyl in crops grown in treated soils
Beynon IK, et al.
Pestic. Sci., 5(4), 443-450 (1974)
Effect of water deficit stress on benzoylprop-ethyl performance and physiological traits of winter wild oat (Avena sterilis subsp. ludoviciana)
Alizade S, et al.
Crop Protection, 137(3), 105292-105292 (2020)
Simultaneous determination of 118 pesticide residues in Chinese teas by gas chromatography-mass spectrometry.
Yang X, et al.
Chemical Papers, 63(1), 39-46 (2009)
Multiresidue method for the determination of residues of 251 pesticides in fruits and vegetables by gas chromatography/mass spectrometry and liquid chromatography with fluorescence detection.
Fillion J, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 83(3), 698- 713 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service