Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

06035

Supelco

Benfuracarb

reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30N2O5S
CAS Number:
Molecular Weight:
410.53
MDL number:
UNSPSC Code:
41116107

grade

reference material

assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

format

neat

storage temp.

−20°C

SMILES string

CCOC(=O)CCN(SN(C)C(=O)Oc1cccc2CC(C)(C)Oc12)C(C)C

InChI

1S/C20H30N2O5S/c1-7-25-17(23)11-12-22(14(2)3)28-21(6)19(24)26-16-10-8-9-15-13-20(4,5)27-18(15)16/h8-10,14H,7,11-13H2,1-6H3

InChI key

FYZBOYWSHKHDMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benfuracarb is a highly toxic, benzofuranyl methylcarbamate insecticide, with lower anti-acetylcholinesterase activity.[1] It is widely used in agriculture[2] to control rootworms and insect pests in rice, corn, beet, maize, citrus, and other agricultural vegetables.[3][1]
This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Application

Benfuracarb may be used as reference material in the following:
  • To investigate its genotoxic effect in human peripheral lymphocytes by using micronucleus (MN) assay.[2]
  • Quantification of the analyte in aromatic herbs[4] and vegetables[5] using chromatography techniques.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

>212.0 °F

flash_point_c

> 100 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leaching potential of carbamates and their metabolites and comparison with triazines
Fava L, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 86(2), 204-208 (2007)
QuEChERS-based method for the determination of carbamate residues in aromatic herbs by UHPLC-MS/MS
Nantia AE, et al.
Food Chemistry, 216(2), 334-341 (2017)
MSPD-GC-MS-MS determination of residues of 15 organic nitrogen-containing pesticides in vegetables
Lin BQ, et al.
Chromatographia, 72(11-12), 1143-1148 (2010)
Evaluation of in vitro genotoxic effects of benfuracarb in human peripheral blood lymphocytes
Erdogmus FS, et al.
Fresenius Environmental Bulletin, 24(3) (2015)
Mutagenic and cytotoxic activities of benfuracarb insecticide.
Eren Y, et al.
Cytotechnology, 68(4), 637- 643 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service