Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05363

Supelco

Etrimfos

reference material

Synonym(s):

O-(6-Ethoxy-2-ethyl-4-pyrimidinyl) O,O-dimethyl thiophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17N2O4PS
CAS Number:
Molecular Weight:
292.29
Beilstein/REAXYS Number:
7136389
MDL number:
UNSPSC Code:
41116107

grade

reference material

form

liquid

shelf life

limited shelf life, expiry date on the label

storage temp.

−20°C

SMILES string

CCOc1cc(OP(=S)(OC)OC)nc(CC)n1

InChI

1S/C10H17N2O4PS/c1-5-8-11-9(15-6-2)7-10(12-8)16-17(18,13-3)14-4/h7H,5-6H2,1-4H3

InChI key

FGIWFCGDPUIBEZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

>212.0 °F

flash_point_c

> 100 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service