Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

03054

Supelco

Tetramethrin

mixture of isomers, reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H25NO4
CAS Number:
Molecular Weight:
331.41
Beilstein/REAXYS Number:
8807938
MDL number:
UNSPSC Code:
41116107

grade

reference material

assay

≥95.0% (sum of isomers, HPLC)

shelf life

limited shelf life, expiry date on the label

composition

cis-Tetramethrin, <25.0% (minor component)

format

neat

storage temp.

−20°C

SMILES string

C\C(C)=C\C1C(C(=O)OCN2C(=O)C3=C(CCCC3)C2=O)C1(C)C

InChI

1S/C19H25NO4/c1-11(2)9-14-15(19(14,3)4)18(23)24-10-20-16(21)12-7-5-6-8-13(12)17(20)22/h9,14-15H,5-8,10H2,1-4H3

InChI key

CXBMCYHAMVGWJQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - STOT SE 2 Inhalation

target_organs

Nervous system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service