Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

00380

Supelco

Glutaric semialdehyde

≥85.0% (GC)

Synonym(s):

5-Oxopentanoic acid, 5-Oxovaleric acid, Glutaraldehydic acid, Glutaric acid semialdehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8O3
CAS Number:
Molecular Weight:
116.12
Beilstein/REAXYS Number:
1747012
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

assay

≥85.0% (GC)

format

neat

storage temp.

−20°C

SMILES string

[H]C(CCCC(O)=O)=O

InChI

1S/C5H8O3/c6-4-2-1-3-5(7)8/h4H,1-3H2,(H,7,8)

InChI key

VBKPPDYGFUZOAJ-UHFFFAOYSA-N

Biochem/physiol Actions

Metabolite of lysine degradation. In the lysine degradation IV pathway, glutarate semialdehyde reacts with NADP+ and H2O to produce glutarate, NADPH, and H+. In this pathway, glutarate semialdehyde is produced by the reaction between 5-aminopentanoate and 2-ketoglutarate, with L-glutamate as a byproduct. The enzyme responsible for this reaction is 5-aminovalerate aminotransferase. In the lysine degradation III pathway, glutarate semialdehyde reacts with NAD+ and H2O to produce glutarate and NADH. In this pathway, glutarate semialdehyde is produced by the reaction between 5-aminopentanoate and 2-ketoglutarate, with L-glutamate as a byproduct. The enzyme responsible for this reaction is 5-aminovalerate aminotransferase.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service