Key Documents
Z769045
Eppendorf® Multipette® M4 pipette
incl. holder, pkg of 1 ea
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
feature
electronic: no
single-channel
packaging
pack of 1 ea
pkg of 1 ea
manufacturer/tradename
Eppendorf® 4982 000.012
volume
1 × 10 μL
compatibility
Combitips advanced® only
General description
The clearly arranged and easy-to-read display guarantees stress-free and intuitive operation. In addition to the displayed dispensing volume, the step counter supports error-free filling, for example plates.
The integrated Sleep function switches off when the Multipette® M4 is not being used, thus reducing energy consumption and saving battery life.
Features:
  • Light and ergonomic
  • Perfectly shaped for large or small hands
  • Weight is well balanced for fatigue-free handling
  • Stress-free work via integrated step counter: dispensing procedures can be continued error-free after an interruption or distraction
  • Central Combitip ejector: fully emptied Combitip can be easily ejected with one hand using the operating lever
Please note: The holder supplied with the Multipette® M4 can be either wall mounted or attached to a carousel stand.
Legal Information
Combitips advanced is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Multipette is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)