Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z768715

Eppendorf® Easypet® 3 electronic pipettor

AC/DC input 100 - 240 V AC

Synonym(s):

liquid handling, pipette, pipette controller, pipetting

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

feature

Lithium-Polymer battery (rechargeable)

manufacturer/tradename

Eppendorf® 4430000018

measuring range

7.1 mL/sec (aspiration speed)

General description

Easypet 3 is a lightweight, well-balanced cordless electronic pipette controller for use with graduated and volumetric glass or plastic serological pipettes from volumes of 0.1-100 mL. Weighing only 160 g with its battery equipped, Easypet 3 has a fast average aspiration speed of 7.1 mL/s.

Although Easypet 3′s average aspiration speed is the second fastest, its lightweight property and the 6-hour operating time, requiring only 3 hours to charge, make it the most convenient pipette controller on the market. Operation while recharging is possible.

Includes:
  • Power supply
  • Wall mounting device
  • Shelf stand
  • 2 membrane filters (unsterile), 0.45 μm pore size

Legal Information

Easypet is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service