Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z765074

KeepIT® 81 plastic freezer box, for batching and storing samples

green

Synonym(s):

freezer containers, storage boxes, storage containers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103000

packaging

pkg of 10 ea

manufacturer/tradename

WHEATON® W651701-G

color

green

Looking for similar products? Visit Product Comparison Guide

General description

KeepIT-81 plastic freezer boxes have six different colors and accommodate 81 external threaded (sizes: 1.2 - 2 mL) cryogenic vials. Openings in bottom facilitate scanning barcodes. Made from Tritan BPA free, shatter resistant resin, these boxes are solvent resistant including DMSO, and withstand cryogenic temperatures.

The standard footprint is compatible with liquid nitrogen storage shelves and freezer drawers.

Legal Information

KeepIT is a registered trademark of Wheaton Industries, Inc.
Tritan is a trademark of Eastman Kodak Company
WHEATON is a registered trademark of Wheaton Industries, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service