Key Documents
Z763047
Eppendorf® Combitips advanced® pipette tips, Sterile grade
maximum volume 10 mL
Synonym(s):
pipetting tips
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
material
orange
sterility
sterile
feature
barrier: no
packaging
pack of 100 ea bag
manufacturer/tradename
Eppendorf® 0030089570
maximum volume
10 mL
General description
The Combitips advanced are optimized for all Repeater models to create the perfect connection!
The new Eppendorf Combitips advanced feature the characteristic Eppendorf 3D design on the upper rim of the Combitip. With 9 volume sizes (0.1 mL to 50 mL) and Eppendorf Quality, Sterile, PCR Clean and Eppendorf Biopur purity grades, you will always find the perfect Combitip for your application.
Sterile grade makes these products applicable for all applications where the absence of living organisms, like microorganisms, could interfere the positive outcome of an experiment. They are also Pyrogen free that ensures absolute absence of endotoxines, which is mandatory for many applications in cell culture, pharmacology and medical research.
Other Notes
Browse all pipette tips
Legal Information
Combitips advanced is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)