Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z762873

Eppendorf® Combitips advanced® pipette tips, PCR clean grade

maximum volume 0.5 mL

Synonym(s):

pipetting tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121600
sterility:
non-sterile
feature:
barrier: no
DNase and RNase free, free of human DNA
packaging:
pack of 100 ea bag
manufacturer/tradename:
Eppendorf® 0030089782

material

purple tip

sterility

non-sterile

feature

barrier: no
DNase and RNase free, free of human DNA

packaging

pack of 100 ea bag

manufacturer/tradename

Eppendorf® 0030089782

technique(s)

PCR: suitable

maximum volume

0.5 mL

Looking for similar products? Visit Product Comparison Guide

General description

The Combitips advanced are optimized for all Repeater models to create the perfect connection!

The new Eppendorf Combitips advanced feature the characteristic Eppendorf 3D design on the upper rim of the Combitip. With 9 volume sizes (0.1 mL to 50 mL) and Eppendorf Quality, Sterile, PCR Clean and Eppendorf Biopur purity grades, you will always find the perfect Combitip for your application.

The PCR clean tubes are free of human DNA, which avoids unspecific signals due to DNA contaminations. They are DNase and RNase free, so that starting material will not be affected by RNA or DNA degrading enzymes. Furthermore, they are also free of any kind of PCR inhibiting agents, which supports highest efficiency of the reaction.

Other Notes

Browse all pipette tips

Legal Information

Combitips advanced is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service