Z762652
Eppendorf® Combitips advanced® pipette tips, Quality grade
maximum volume 1.0 mL
All Photos(1)
About This Item
UNSPSC Code:
41121600
Recommended Products
material
yellow tip
sterility
non-sterile
feature
barrier: no
packaging
pack of 100 ea
manufacturer/tradename
Eppendorf® 0030089430
maximum volume
1.0 mL
General description
The Combitips advanced are optimized for all Repeater models to create the perfect connection!
The new Eppendorf Combitips advanced feature the characteristic Eppendorf 3D design on the upper rim of the Combitip. With 9 volume sizes (0.1 mL to 50 mL) and Eppendorf Quality, Sterile, PCR Clean and Eppendorf Biopur purity grades, you will always find the perfect Combitip for your application.
With the redesigned dispenser box for the Eppendorf Quality tips, the extricable chute makes Combitips advanced always within reach. Portioning in 4 bags (25 pcs. each) ensures dust-free storage.
The new Eppendorf Combitips advanced feature the characteristic Eppendorf 3D design on the upper rim of the Combitip. With 9 volume sizes (0.1 mL to 50 mL) and Eppendorf Quality, Sterile, PCR Clean and Eppendorf Biopur purity grades, you will always find the perfect Combitip for your application.
With the redesigned dispenser box for the Eppendorf Quality tips, the extricable chute makes Combitips advanced always within reach. Portioning in 4 bags (25 pcs. each) ensures dust-free storage.
Legal Information
Combitips advanced is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)