Key Documents
Z761214
Digital video camera for Jenco stereo microscopes
5 Megapixel, USB 2.0 cable with adapter and image capture software
About This Item
UNSPSC Code:
41191506
NACRES:
NB.79
manufacturer/tradename
Jenco 57-5M
General description
Jenco offers three types of camera systems for documenting resolved microscopic images: digital cameras, digital videos or analog video cameras.
Each system is fully integrated with:
  • Software for image analysis
  • Optical adapters to obtain the image in the correct focal plane
  • Cables for connection to computers, monitors or projectors
Jenco Image is a software tool for microscope image measurement. It captures or imports and archives database images and allows image annotation, E-mail links, or report generation. It can be used to measure point to point or multi-pointed lengths/distances and/or area of: lines, angles, circles, rectangles and ellipses in the captured image with calipers in metric, English or pixels.
The system can also calculate a color area and percentage of color relative to the whole. The system can be calibrated to scale/magnification so the result of the measurements and virtual values will keep a 1:1 relation. All results can be saved in the computer for further research.
  • Point-to-point linear measurement
  • Radius and diameter on ellipse
  • Area and perimeter for ellipse or rectangle
  • Length of segmented lines
  • Area and perimeter of irregular shapes
  • Angle measurements
  • Arc/Ellipse/Circle measurements including definition of center point, chord lengths, sweep angles, and radii
Legal Information
Jenco is a trademark of Jenco International, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)