Key Documents
Z756962
Burco Drosophila fly anesthetizer
polyethylene, pack of 1 ea
About This Item
UNSPSC Code:
41100000
NACRES:
NB.22
material
polyethylene
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F14300-0000
General description
Rapid action, optimally sized chamber for anesthetizing Drosophila. It is ideal for classroom and laboratory use or demonstration. The chamber can be used with the cap seat of a regular 1/2 pint milk bottle and can be altered to be compatible with other fly containers. Detailed instructions included.
Legal Information
Burco is a trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)