Key Documents
Z756881
Scienceware® antibody saver tray
L × W 165 mm × 165 mm, pack of 5 ea
Synonym(s):
incubation trays
About This Item
UNSPSC Code:
41122800
packaging
pack of 5 ea
manufacturer/tradename
Bel-Art F45100-0002
L × W
165 mm × 165 mm
General description
The antibody saver trays help minimize the use of expensive antibodies when incubating filters in the Western Blot process by using an optimally sized tray.
  • Includes attached lid to protect against spillage
  • Trays are stackable, reclosable and reusable (1.5 cm deep)
  • Molded of polypropylene
  • Autoclavable at 121ºC (250ºF)
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)